Learn More
Pyrex™ Borosilicate Glass Flat Bottom Evaporating Dishes
Product Code. 15127193
Click to view available options
Capacity (Metric):
15 mL
45 mL
90 mL
170 mL
320 mL
600 mL
Diameter (Metric):
50 mm
60 mm
80 mm
95 mm
115 mm
140 mm
Height (Metric):
25 mm
30 mm
45 mm
55 mm
65 mm
80 mm
Unit Size:
Each
Pack of 10
Product Code. 15127193
Brand: Pyrex 1450/06D
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from Pyrex™ borosilicate glass and complies to DIN 12336 standards. Pyrex™ Borosilicate Glass Flat Bottom Evaporating Dishes are highly resistant to chemical erosion and thermal shock. Flat base with curved sides and spout provide easy handling and helps to prevent spillage.
  • Complies to DIN 12336
  • Manufactured from Pyrex™ borosilicate glass
  • High resistance to chemical erosion and thermal shock
  • Flat base
  • Curved sides with spout
TRUSTED_SUSTAINABILITY
Specifications
Bottom Shape Flat
Capacity (Metric) 170 mL
Material Borosilicate Glass
Shape Round
Type Evaporating Dish
Height (Metric) 55 mm
Diameter (Metric) 95 mm