Learn More
Pyrex™ Borosilicate Glass Erlenmeyer Flask with Quickfit Ground Glass Socket
Description
Grow cultures and store other laboratory reagents in Glass Erlenmeyer flasks. The Pyrex™ borosilicate glass Erlenmeyer flask with QuickFit™ glass socket is sturdy and comes in a variety of sizes.
- Complies to ISO 4797
- Conical flasks with highest quality Quickfit™ ground glass socket
- Borosilicate glass for corrosion free performance and high resistance to thermal shock
- Erlenmeyer flasks with high quality Quickfit™ ground glass socket
- Manufactured from Pyrex™ borosilicate glass for highest resistance to thermal shock and chemical corrosion
- White enamel graduations and marking spot
Specifications
Specifications
| Certifications/Compliance | DIN 12387 |
| Capacity (Metric) | 100 mL |
| Material | Borosilicate Glass |
| Graduations | Yes |
| Height (Metric) | 110 mm |
| Shape | Conical |
| Standard Taper Joint | 29/32 |
| Writing Surface | Yes |
| Graduated | Yes |
| Diameter (Metric) Outer | 64 mm |