Learn More
Pyrex™ Borosilicate Glass Cylindrical Conical Flask
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
Height (Metric):
70 mm
97 mm
130 mm
Diameter (Metric) Outer:
45 mm
60 mm
69 mm
90 mm
Unit Size:
Pack of 10
Pack of 20
Pack of 50
Description
- Manufactured from Pyrex™ borosilicate glass for effective resistance to thermal shock and chemical corrosion
- White enamel marking spot
- Autoclavable
Specifications
Specifications
| Diameter (Metric) Outer Neck | 30 mm |
| Capacity (Metric) | 125 mL |
| Material | Borosilicate Glass |
| Height (Metric) | 97 mm |
| Shape | Conical, Cylindrical |
| Writing Surface | Yes |
| Autoclavable | Autoclavable |
| Diameter (Metric) Outer | 60 mm |