Learn More
Pyrex™ Borosilicate Glass Class A Volumetric Flask
Click to view available options
Capacity (Metric):
1 mL
2 mL
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
5 L
Closure Size:
10/19
12/21
14/23
19/26
24/29
29/32
34/35
7/16
Height (Metric):
65 mm
70 mm
90 mm
110 mm
140 mm
170 mm
210 mm
220 mm
260 mm
300 mm
370 mm
475 mm
Tolerance:
±0.02 mL
±0.03 mL
±0.05 mL
±0.08 mL
±0.1 mL
±0.12 mL
±0.2 mL
±0.3 mL
±0.5 mL
±1 mL
Dimensione della confezione:
Each
Pack of 2
Pack of 5
Descrizione
Manufactured from chemically resistant Pyrex™ Borosilicate Glass. Pyrex™ Borosilicate Glass Class A Volumetric Flask includes white enamel inscriptions for easy volume assessment and a polyethylene stopper. Complies with ISO 1042 and DIN 12664, is calibrated to the tolerance required by the United States Pharmacopoeia (USP,) and Conformity 'H' certified.
- Complies with ISO 1042 and DIN 12664
- Calibrated to the tolerance required by the United States Pharmacopoeia (USP) which is tighter than those of ISO/DIN standards
- Each flask is laser etched with an individual serial number batch code and date of manufacture
- Batch certificate available to download online
- Conformity 'H' certified
- Manufactured from chemically resistant Pyrex™ glass
- Inscriptions in white enamel
- Polyethylene stopper
Specifica
Specifica
Certifications/Compliance | ISO 1042, DIN 12664, USP, Conformity 'H' certified |
Closure Material | Polyethylene |
Description | White enamel inscriptions |
Capacity (Metric) | 1000 mL |
Material | Borosilicate Glass |
Height (Metric) | 300 mm |
Tolerance | ±0.3 mL |
Graduated | Yes |
Class | A |
Closure Size | 24/29 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Pyrex™ Borosilicate Glass Class A Volumetric Flask >
Individuate un'opportunitĂ di miglioramento?Condividi una correzione di contenuto