Learn More
Pyrex™ Aspirator Bottle with Ground Glass Side Socket
Product Code. 11337884
Change view
Click to view available options
Capacity (Metric):
10,000 mL
20,000 mL
2000 mL
5000 mL
Unit Size:
Each
Pack of 5
Product Code. Capacity (Metric) unitSize
11337884 2000 mL Each
11357884 10,000 mL Each
11377884 20,000 mL Each
13421909 2000 mL Pack of 5
11347884 5000 mL Each
5 options
Product Code. 11337884
Brand: Pyrex 1541/12D
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from borosilicate glass
  • Ground side socket (size 29/32) to accept stopcock assemblies
  • Unground top neck to accept rubber stoppers
Compliance
ISO 4796
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 2000 mL
Diameter (Metric) Inner Neck 28 mm
Material Borosilicate Glass
Height (Metric) 248 mm
Product Type Aspirator Bottle
Diameter (Metric) 138 mm