Learn More
Pyraclonil, Dr. Ehrenstorfer
Product Code. 30310727
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 30310727
Brand: Dr. Ehrenstorfer DREC16593000
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Pyraclonil
CAS 158353-15-2
Analyte or Component Names Pyraclonil
Shipping Condition Room Temperature
Molecular Formula C15 H15 Cl N6
Certifications/Compliance ISO 17034
Synonym 1H-Pyrazole-4-carbonitrile, 1-(3-chloro-4,5,6,7-tetrahydropyrazolo[1,5-a]pyridin-2-yl)-5-(methyl-2-propyn-1-ylamino)- (ACI), 1-(3-Chloro-4,5,6,7-tetrahydropyrazolo[1,5-a]pyridin-2-yl)-5-(methyl-2-propyn-1-ylamino)-1H-pyrazole-4-carbonitrile (ACI), 1H-Pyrazole-4-carbonitrile, 1-(3-chloro-4,5,6,7-tetrahydropyrazolo[1,5-a]pyridin-2-yl)-5-(methyl-2-propynylamino)- (9CI), Pyrazolo[1,5-a]pyridine, 1H-pyrazole-4-carbonitrile deriv. (ZCI), AEB 391, Diazolinonitrile, EP 913, HSA 961, Pyraclon, Pyraclonil, Pyrazogyl
InChI Formula InChI=1S/C15H15ClN6/c1-3-7-20(2)15-11(9-17)10-18-22(15)14-13(16)12-6-4-5-8-21(12)19-14/h1,10H,4-8H2,2H3
Solution Type Neat
SMILES CN(CC#C)c1c(cnn1c2nn3CCCCc3c2Cl)C#N
IUPAC Name 1-(3-chloro-4,5,6,7-tetrahydropyrazolo[1,5-a]pyridin-2-yl)-5-[methyl(prop-2-ynyl)amino]pyrazole-4-carbonitrile
Industry Type Environmental Testing
Molecular Weight (g/mol) 314.77
Quantity 10 mg
Formula Weight 314.1047 g/mol