missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyr10, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
Pyr10 is a pyrazole derivative and a selective TRP cation 3 (TRPC3) inhibitor. Pyr10 inhibits Ca2+ influx in carbachol-stimulated TRPC3-transfected HEK293 cells with an IC50 of 0.72 μM (IC50 of 13.08 μM for store operated Ca2+ entry in BRL-2H3 cells). Pyr10 has the ability to distinguish between receptor-operated TRPC3 and native stromal interaction molecule 1 (STIM1)/Orai1 channels.
Specifications
Specifications
| Chemical Name or Material | Pyr10 |
| CAS | 1315323-00-2 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C18H13F6N3O2S |
| Quantity | 5 mg |
| Solubility Information | DMSO : 100 mg/mL (222.53 mM; Need ultrasonic) |
| SMILES | O=S(C1=CC=C(C)C=C1)(NC2=CC=C(N3N=C(C(F)(F)F)C=C3C(F)(F)F)C=C2)=O |
| Molecular Weight (g/mol) | 449.37 |
| Formula Weight | 449.37 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction