Learn More
Karl Hecht™ Pycnometers According to Gay-Lussac
Product Code. 10758102
Change view
Click to view available options
Capacity (Metric):
5 mL
50 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10758102 5 mL Each
10217341 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10758102 Supplier Karl Hecht™ Supplier No. 42571008
  • ONLINE EXCLUSIVE PRICE
    984.06 NOK
    630.64 NOK / Pack of 1
    Save 353.42 NOK
    35% Off
 Restricted Item
Estimated Shipment: 23-12-2026
For the measurement of liquid density in the laboratory.
  • With ground-in stopper
  • Precisely adjusted
  • Available in different capacities
Specifications
Type Pycnometer acc. to Gay-Lussac
Capacity (Metric) 5 mL
For Use With (Application) Measuring liquid density
Description With ground-in stopper