Learn More
Karl Hecht™ Pycnometers According to Gay-Lussac
Product Code. 10217341
Change view
Click to view available options
Capacity (Metric):
5 mL
50 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10217341 50 mL Each
10758102 5 mL Each
2 options
Product Code. 10217341
Brand: Karl Hecht™ 42571032
Only null left
Add to basket
Add to basket
For the measurement of liquid density in the laboratory.
  • With ground-in stopper
  • Precisely adjusted
  • Available in different capacities
Specifications
Type Pycnometer acc. to Gay-Lussac
Capacity (Metric) 50 mL
For Use With (Application) Measuring liquid density
Description With ground-in stopper