Learn More
PX-12, MedChemExpress
Product Code. 16491977
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16491977
Brand: Medchem Express HY13734
MedChemExpress PX-12(IV-2) is an irreversible inhibitor of Thioredoxin-1 (Trx-1); inhibits the growth of MCF-7 and HT-29 cells with IC50 values of 1.9 and 2.9 μM, respectively.
PX-12(IV-2) is an irreversible inhibitor of Thioredoxin-1 (Trx-1)
Specifications
Chemical Name or Material PX-12
Physical Form Solid
CAS 141400-58-0
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C7H12N2S2
For Use With (Application) Cancer-programmed cell death
Synonym IV-2
Solubility Information Ethanol : 50 mg/mL (265.52 mM; Need ultrasonic) ∣DMSO : ≥ 44.7 mg/mL (237.37 mM)
SMILES CCC(SSC1=NC=CN1)C
Molecular Weight (g/mol) 188.31
Formula Weight 188.31
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.