Learn More
Masterflex™ PVDF Reducing Nipple
Description
- Use with our rigid tubing for high-pressure applications
- Good chemical resistance
- Rated to 200psi at 77°F (25°C)
- Usable from -40 to +200°F (-40 to +93°C)
Specifications
Specifications
| Description | NPT(M) Connection |
| Autoclavable | Not Autoclavable |
| Material | PVDF |
| Packaging Quantity | 6 Fittings |
| Port Size | 1/2 x 1/8 in. NPT(M) |
| Sterilization Method | Ethylene Oxide |
| Thread Size | 1/2 x 1/8 in. NPT(M) |
| Product Type | Reducing Nipple |