Learn More
Cole-Parmer™ PVDF Manifold, 6 port
Product Code. 11705288
Change view
Click to view available options
Tubing Size:
1/2 in. x 3/8 in.
3/8 in. x 1/4 in.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tubing Size unitSize
11705288 3/8 in. x 1/4 in. Each
11715288 1/2 in. x 3/8 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11705288 Supplier Cole-Parmer™ Supplier No. 3152345
Only null left
Add to basket
Add to basket
Expand your flow and line capacity with these manifolds. Choose the number of ports and connections to suit your needs.
  • Constructed of durable PVDF.
  • Ideal for applications requiring excellent chemical compatibly.
  • Available in a variety of configurations
Specifications
Material PVDF
Overall Length (English) 5 7/8 in.
Overall Length (Metric) 14.93 cm
Min. Operating Temperature (English) -65°F
Min. Operating Temperature (Metric) -54°C
Tubing Size 3/8 in. x 1/4 in.
Inlet 3/8 in. NPT(F)
Outlet Port 1/4 in. NPT(F)
Width (Metric) 3.18 cm
Width (English) 1.25 in.
Height (Metric) 3.18 cm
Height (English) 1.25 in.
Maximum Pressure 110 psi
Max. Operating Temperature (English) 230°F
Max. Operating Temperature (Metric) 110°C
Product Type Manifold