Learn More
Cole-Parmer™ PVDF Manifold, 2 port
Product Code. 11745278
Change view
Click to view available options
Tubing Size:
1/2 in. x 3/8 in.
1/4 in. x 1/8 in.
3/8 in. x 1/4 in.
Unit Size:
Each
Product Code. Tubing Size unitSize
11745278 3/8 in. x 1/4 in. Each
11735278 1/4 in. x 1/8 in. Each
11755278 1/2 in. x 3/8 in. Each
3 options
Product Code. 11745278
Brand: Cole-Parmer™ 3152333
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Expand your flow and line capacity with these manifolds. Choose the number of ports and connections to suit your needs.
  • Constructed of durable PVDF.
  • Ideal for applications requiring excellent chemical compatibly.
  • Available in a variety of configurations
Specifications
Material PVDF
Overall Length (English) 2 3/8 in.
Overall Length (Metric) 6.03 cm
Min. Operating Temperature (English) -65°F
Min. Operating Temperature (Metric) -54°C
Tubing Size 3/8 in. x 1/4 in.
Inlet 3/8 in. NPT(F)
Outlet Port 1/4 in. NPT(F)
Width (Metric) 3.18 cm
Width (English) 1.25 in.
Height (Metric) 3.18 cm
Height (English) 1.25 in.
Maximum Pressure 110 psi
Max. Operating Temperature (English) 230°F
Max. Operating Temperature (Metric) 110°C
Product Type Manifold