Learn More
Cole-Parmer™ PVDF Bulkhead Compression Union
Click to view available options
Port Size:
1/2 in.
3/8 in.
5/16 in.
Unit Size:
Each
Description
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
- Very good chemical resistance
- Rated to 220psi maximum at 77°F ( 25°C)
- Usable from 32 to 212°F (0 to 100°C)
Specifications
Specifications
| Description | Compression Connection |
| Autoclavable | Autoclavable |
| Material | PVDF |
| Overall Length (English) | 2 21/32 in. |
| Tubing Size | 3/8 in. |
| Outlet Port | 3/8 in. |
| Port Size | 3/8 in. |
| Sterilization Method | Autoclave or Ethylene Oxide |
| Width (English) | 13/16 in. |
| Product Type | Bulkhead Compression Union |