Learn More
Deutsch Neumann™ Vacuum and Pressure Bulbs
Click to view available options
:
Vacuum Pressure Bulb
Description
- Reddish-brown in color
- With rigid connectors
- Non-return valves ensure unidirectional flow
Specifications
Specifications
| Color | Brown |
| Material | PVC |
| Type | Vacuum Pressure Bulb |
| For Use With (Equipment) | For orsatapparat |