Learn More
Buerkle™ PVC Wide Mouth Containers
Product Code. 10630704 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
200 mL
300 mL
500 mL
750 mL
1000 mL
2000 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10630704 50 mL Each
15326438 100 mL Each
15455564 200 mL Each
11828232 300 mL Each
11838232 500 mL Each
10156743 750 mL Each
11812633 1000 mL Each
11822633 2000 mL Each
8 options
Product Code. 10630704
Brand: Buerkle™ 03550050
Only null left
Add to basket
Add to basket
Manufactured with highly transparent PVC for optimum visibility and impact resistance. Buerkle™ PVC Wide Mouth Containers provide a wide opening for easy and safe filling.
  • Polypropylene black screw lid with polyethylene seal
  • Suitable for liquid, powder or granules, capsules, dyes
  • Use for transport of samples and specimens
  • Square shape optimizes storage space
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 50 mL
Color Clear
Dimensions (L x W x H) 38 x 38 x 63 mm
Diameter (Metric) Inner Neck 23 mm
For Use With (Application) For Liquids, Powders or Granules Such as Tablets, Capsules, Dyes, Shipments of Samples, Specimens Etc.
Material PVC
Length (Metric) 38mm
Neck ID (Metric) 23 mm
Quantity 1 Ea.
Transparency Yes
Width (Metric) 38mm
Closure Color Black
Closure Included Yes
Mouth Wide
Shape Square
Height (Metric) 63 mm
Product Type Jar
Closure Material Polypropylene
Closure Type Screw Cap