Learn More
Buerkle™ PVC Wide Mouth Containers
Product Code. 10156743 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
200 mL
300 mL
500 mL
750 mL
1000 mL
2000 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10156743 750 mL Each
15326438 100 mL Each
15455564 200 mL Each
10630704 50 mL Each
11828232 300 mL Each
11838232 500 mL Each
11812633 1000 mL Each
11822633 2000 mL Each
8 options
Product Code. 10156743
Brand: Buerkle™ 03550750
Only null left
Add to basket
Add to basket
Manufactured with highly transparent PVC for optimum visibility and impact resistance. Buerkle™ PVC Wide Mouth Containers provide a wide opening for easy and safe filling.
  • Polypropylene black screw lid with polyethylene seal
  • Suitable for liquid, powder or granules, capsules, dyes
  • Use for transport of samples and specimens
  • Square shape optimizes storage space
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 750 mL
Color Transparent
Dimensions (L x W x H) 92 x 92 x 120 mm
Diameter (Metric) Inner Neck 69 mm
Material PVC
Length (Metric) 92mm
Width (Metric) 92mm
Closure Color Black
Closure Included Yes
Mouth Narrow
Shape Square
Height (Metric) 120 mm
Closure Liner Polyethylene
Closure Material Polypropylene
Closure Type Screw Cap