Learn More
Cole-Parmer™ PVC Stopcocks with Flow Indication
Product Code. 11809442
Change view
Click to view available options
Connectors:
Hose x Hose
NPT(M) x Hose
Fits Tubing ID:
0.25 to 0.5 in.
0.5 to 5/8 in.
Unit Size:
Each
Product Code. Connectors Fits Tubing ID unitSize
11809442 NPT(M) x Hose 0.5 to 5/8 in. Each
11809452 Hose x Hose 0.25 to 0.5 in. Each
2 options
Product Code. 11809442
Brand: Cole-Parmer™ 0622554
Only null left
Add to basket
Add to basket
Eliminate guess work with these stopcocks with flow indication. Cole Parmer™ PVC Stopcocks with Flow Indication feature PVC bodies, EPDM seals and seats. Flow indicator on handle.
  • Maximum pressure is 150psi at the maximum temperature of 49°C
  • Maximum vacuum is 29.9'' Hg
Specifications
Material PVC
Pressure 150 psi
Fits Tubing ID 0.5 to 5/8 in.
No. per Pack 1 Each
No. per Case 1 Each
Connectors NPT(M) x Hose