Learn More
Buerkle™ PVC Standard Bung
Laboratory standard bung made of PVC with good chemical resistance against acids and alkalis.
Brand:  Buerkle™ 9763-1924
41.25 NOK valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 17250644
  • 43.85 NOK / Each
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
Alert:
Attention! Make sure to pay attention to the “List of Chemical Resistance“!
Description
Description
  • PVC, soft, red
Specifications
Specifications
Red
19 mm
PVC
25 mm
24 mm