Learn More
Clark Scientific™ PVC Liners
Product Code. 12908847
Change view
Click to view available options
Dimensions (L x W):
470 x 360 mm
Unit Size:
Pack of 10
Product Code. Dimensions (L x W) unitSize
12908847 470 x 360 mm Pack of 10
1 options
Product Code. 12908847
Brand: Clark Scientific™ SSmall
Only null left
Add to basket
Add to basket
Colorless PVC liners for rigid PVC spill trays that are ideal in most laboratory environments.
  • Colorless PVC liners
  • Liners for rigid PVC spill trays
  • Ideal for use in laboratories
Specifications
Product Type Liner
Material PVC
Length (Metric) 470 mm
Width (Metric) 360 mm
Dimensions (L x W) 470 x 360 mm
Backing Material PVC
Quantity 10Pack