Learn More
- Home
- Products
- Tubing
- Rubber and Silicone Tubing
- Cole-Parmer PVC Lab and Vacuum Tubing
Cole-Parmer PVC Lab and Vacuum Tubing
Product Code. 30053670 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (English) Inner:
1 in.
1/16 in.
1/2 in.
1/4 in.
1/5 in.
1/8 in.
15/64 in.
3/16 in.
3/32 in.
3/4 in.
3/8 in.
5/16 in.
5/32 in.
5/64 in.
Diameter (English) Outer:
1 in.
1-1/4 in.
1-1/8 in.
1/2 in.
1/4 in.
1/5 in.
1/8 in.
11/32 in.
15/32 in.
15/64 in.
3/16 in.
3/4 in.
3/8 in.
5/16 in.
5/32 in.
5/8 in.
7/16 in.
7/8 in.
9/16 in.
Unit Size:
15m
| Product Code. | Diameter (English) Inner | Diameter (English) Outer | unitSize |
|---|---|---|---|
| 30053670 | 5/16 in. | 15/32 in. | 15m |
| 30054031 | 3/8 in. | 1/2 in. | 15m |
| 30053338 | 1/16 in. | 1/8 in. | 15m |
| 30053611 | 1/4 in. | 5/8 in. | 15m |
| 30053704 | 1/2 in. | 5/8 in. | 15m |
| 30053850 | 1/2 in. | 3/4 in. | 15m |
| 30053084 | 3/32 in. | 5/32 in. | 15m |
| 30053748 | 3/8 in. | 7/8 in. | 15m |
| 30054239 | 5/64 in. | 5/32 in. | 15m |
| 30054309 | 1/2 in. | 1-1/8 in. | 15m |
| 30053306 | 3/8 in. | 9/16 in. | 15m |
| 30053415 | 1/8 in. | 1/4 in. | 15m |
| 30053287 | 3/4 in. | 1 in. | 15m |
| 30053926 | 1 in. | 1-1/4 in. | 15m |
| 30053701 | 1/4 in. | 3/8 in. | 15m |
| 30052936 | 5/16 in. | 1/2 in. | 15m |
| 30053886 | 5/16 in. | 7/16 in. | 15m |
| 30053433 | 3/16 in. | 9/16 in. | 15m |
| 30053381 | 1/8 in. | 3/16 in. | 15m |
| 30053136 | 3/8 in. | 5/8 in. | 15m |
| 30053449 | 5/32 in. | 15/64 in. | 15m |
| 30053173 | 1/8 in. | 1/5 in. | 15m |
| 30054048 | 1/5 in. | 5/16 in. | 15m |
| 30054189 | 15/64 in. | 11/32 in. | 15m |
| 30052944 | 1/4 in. | 7/16 in. | 15m |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
Versatile applications including peristaltic pumps, analytical instruments, incubators, gas, and drain lines. Glass-smooth inner bore design for easy clean-ups and changeovers, enhancing workflow efficiency. Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Specifications
Specifications
| Diameter (English) Inner | 5/16 in. |
| Material | PVC |
| Diameter (English) Outer | 15/32 in. |
| Overall Length (English) | 50 ft. |
| Color | Translucent |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.