Learn More
Brady™ PVC ISO First Aid Eyewash Sign
Specifications
Specifications
| Certifications/Compliance | ISO 7010 |
| Alternate Part Number | PIC E011-203x203-VPVC-CRD/1 |
| Material | PVC |
| Quantity | 1 Ea. |
| For Use With (Application) | Indoor & Outdoor |
| Height (Metric) | 203 mm |
| Thickness | 1.591 mm |
| Width (Metric) | 203.00 mm |
| Temperature Range (Metric) | -20 °C-70 °C |
| Shape | Square |
| Show More |