Learn More
Cole-Parmer™ Digi-Sense™ Type-T PVC-Insulated Probes with Epoxy-Coated Tip
Product Code. 11731349 Shop All Cole-Parmer Products
Change view
Click to view available options
Temperature (Metric):
-200°C to +105°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature (Metric) Unit Size
11731349 -200°C to +105°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11731349 Supplier Cole-Parmer™ Supplier No. 0846606
  • ONLINE EXCLUSIVE PRICE
    419.00 NOK
    394.18 NOK / Pack of 1
    Save 24.82 NOK
    5% Off
View all promotions
Estimated Shipment: 29-01-2027
Use these probes for applications requiring an isolated junction. Digi-Sense™ Type-T PVC-Insulated Probes with Epoxy-Coated Tip are ideal for short term immersion.
Each probe comes with a color coded ANSI mini connector: type J-black, type K-yellow, type T-blue and type E-purple. Response Time: 2 seconds or greater.
  • Exposed-junction probes are not recommended for use with aqueous solutions
  • Do not subject type J exposed-junction probes to moisture as they will rust
Specifications
Certifications/Compliance ANSI
Temperature (Metric) -200°C to +105°C
Type Thermocouple Probe
For Use With (Application) Used for Short Term Immersion
Length (Metric) 20.3 cm