Learn More
OHAUS™ PVC Flask Clamp
Shop All Ohaus Products
Click to view available options
For Use With (Equipment):
1000 mL Flask
125 mL Flask
2000 mL Flask
250 mL Flask
500 mL Flask
Unit Size:
Each
Description
- Constructed of PVC
- For use with flasks
- Designed to securely hold flask in place
Specifications
Specifications
| Quantity | 1 |
| Type | Flask Clamp |
| Material | PVC |
| For Use With (Equipment) | 2000 mL Flask |
| Description | Flask Clamp PVC 2000mL |
| Color | Purple |
| Jaw Type | Four Interlocking |
| Diameter (Metric) Inside | 6.68 |