Description
- Easily moved for cleaning
- Non-skid backing ensures frame stays in place
- Fits all PURUS™ EnviroTack™ mats
- Strong and Durable
- Non-Slip made from Polymer Coated polyester textile
- Naturally antimicrobial, mold and mildew resistant
Specifications
Specifications
| Dimensions (L x W) | 508 x 1194 mm |
| For Use With (Application) | For use in Pharmaceutical, Biotech, Semiconductor, Cleanrooms, and Industrial Applications |
| For Use With (Equipment) | 457 x 1143 mm PURUS™ Biohybrid™ Mats |
| Thickness (Metric) | 3.175 mm |
| Color | White |
| Material | Polystyrene |