Learn More
PURUS™ Frames for EnviroTack™ Contamination Control Mats
Product Code. 15342002 Shop All Purus Products
Change view
Click to view available options
Dimensions (L x W):
508 x 965 mm
508 x 1194 mm
711 x 1194 mm
965 x 1194 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
15342002 508 x 965 mm Each
15352002 508 x 1194 mm Each
15157162 711 x 1194 mm Each
15382002 965 x 1194 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15342002 Supplier PURUS™ Supplier No. PF 2038 01 W 00
  • ONLINE EXCLUSIVE PRICE
    1241.00 NOK
    1127.26 NOK / Pack of 1
    Save 113.74 NOK
    9% Off
View all promotions
Estimated Shipment: 26-10-2026
Add to basket
Add to basket
Designed to be used with the PURUS™ EnviroTack™ mat to create a system that is easy to use in any critical area where direct adhesion of mats to a flooring surface is not allowed.
  • Easily moved for cleaning
  • Non-skid backing ensures frame stays in place
  • Fits all PURUS™ EnviroTack™ mats
  • Strong and Durable
  • Non-Slip made from Polymer Coated polyester textile
  • Naturally antimicrobial, mold and mildew resistant
Specifications
Dimensions (L x W) 508 x 965 mm
For Use With (Application) For use in Pharmaceutical, Biotech, Semiconductor, Cleanrooms, and Industrial Applications
For Use With (Equipment) 457 x 914 mm PURUS™ Biohybrid™ Mats
Thickness (Metric) 3.175 mm
Color White
Material Polystyrene