Learn More
Purgeable Aromatics For Gasoline IdentifiCation, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| 1,2-Dichlorobenzene | 95-50-1 | 2000μg/mL |
| 1,3-Dichlorobenzene | 541-73-1 | 2000μg/mL |
| 1,4-Dichlorobenzene | 106-46-7 | 2000μg/mL |
| Benzene | 71-43-2 | 2000μg/mL |
| Chlorobenzene | 108-90-7 | 2000μg/mL |
| Ethylbenzene | 100-41-4 | 2000μg/mL |
| Methyl-tert-butyl ether | 1634-04-4 | 2000μg/mL |
| m-Xylene | 108-38-3 | 2000μg/mL |
| o-Xylene | 95-47-6 | 2000μg/mL |
| p-Xylene | 106-42-3 | 2000μg/mL |
| Toluene | 108-88-3 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | Purgeable Aromatics |
| Packaging | 2mL amber prescored glass ampule |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
| Methods | US EPA Method 602 |
| Molecular Formula | CH4O |
| MDL Number | MFCD00004595 |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| SMILES | CO |
| IUPAC Name | methanol |
| Show More |
Safety and Handling
Product Identifier
- Purgeable Aromatics for Gasoline Identification
- Danger
- Acute toxicity Category 3
- Carcinogenicity Category 1A
- Flammable liquid Category 2
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H361-Suspected of damaging fertility or the unborn child.
- H370-Causes damage to organs.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : 1,4-dichlorobenzene,benzene, toluene, ethylbenzene, methanol
Product intended for laboratory use only