Learn More
Labcon™ PurePlus™ Cap Strips for 1.2 mL Sample Library Tubes
Description
- For use with PurePlus™ 1.2 mL Sample Library Tubes
Specifications
Specifications
| Color | Natural |
| Format | Strips of 12 |
| Quantity | 80 Tubes/Pk. |
| Sterility | Non-sterile |
| For Use With (Equipment) | PurePlus™ 1.2 mL Sample Library Tubes |