Learn More
Keystone Cleanroom Products PureClean Self-Sealing 1421B Tyvek™ Autoclave Bag with Clear Film Side and Steam Indicator, 10 to 15 in. wide
GREENER_CHOICE
Product Code. 30042302 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Size:
10 x 15
10 x 18
10 x 20
10 x 22
10 x 24
10 x 30
10 x 36
10 x 48
10 x 56
10 x 62
12 x 12
12 x 14
12 x 16
12 x 18
12 x 20
12 x 22
12 x 24
12 x 28
12 x 30
12 x 32
12 x 34
12 x 36
12 x 40
12 x 42
12 x 46
12 x 48
14 x 14
14 x 16
14 x 18
14 x 20
14 x 24
14 x 26
14 x 36
14 x 38
14 x 40
14 x 48
15 x 26
15 x 43
Unit Size:
Pack of 250
Product Code. Size unitSize
30042302 14 x 40 Pack of 250
30042304 14 x 24 Pack of 250
30042225 14 x 20 Pack of 250
30042317 14 x 18 Pack of 250
30042243 14 x 16 Pack of 250
30042278 14 x 14 Pack of 250
30042299 12 x 48 Pack of 250
30042280 12 x 46 Pack of 250
30042203 12 x 42 Pack of 250
30042179 12 x 40 Pack of 250
30042244 12 x 36 Pack of 250
30042321 12 x 34 Pack of 250
30042207 12 x 32 Pack of 250
30042205 12 x 30 Pack of 250
30042277 12 x 28 Pack of 250
30042315 12 x 24 Pack of 250
30042231 12 x 22 Pack of 250
30042188 12 x 20 Pack of 250
30042189 12 x 18 Pack of 250
30042257 12 x 16 Pack of 250
30042255 12 x 14 Pack of 250
30042216 12 x 12 Pack of 250
30042248 10 x 62 Pack of 250
30042191 10 x 56 Pack of 250
30042251 10 x 48 Pack of 250
30042270 10 x 36 Pack of 250
30042220 10 x 30 Pack of 250
30042228 10 x 24 Pack of 250
30042212 10 x 22 Pack of 250
30042246 10 x 20 Pack of 250
30042184 10 x 18 Pack of 250
30042206 10 x 15 Pack of 250
30042201 15 x 43 Pack of 250
30042263 15 x 26 Pack of 250
30042208 14 x 48 Pack of 250
30042281 14 x 38 Pack of 250
30042215 14 x 36 Pack of 250
30042193 14 x 26 Pack of 250
38 options
Product Code. 30042302
Brand: Keystone Cleanroom Products 6STKRP1440SS
Only null left
Add to basket
Add to basket
PureClean™ Self-Sealing Autoclave Bags are made from a transparent film and 1421B DuPont™ Tyvek™, which is a new grade of uncoated, untreated, flexible Tyvek™ material specifically designed for pharmaceutical use in steam sterilization applications.
  • Uncoated Tyvek™ eliminates potential of residue deposits on equipment, post sterilization
  • Clear Side enables visual confirmation of dryness for sterilized parts and equipment, which is required by EU GMP Annex 1
  • Convenient Self-Seal Closure eliminates the need for autoclave tape or the validation and use of a heat sealer
  • Fast and consistent for equipment preparation area
  • Resists tearing and punctures
  • Superior microbial barrier
  • Manufactured and packaged in a certified ISO 6 cleanroom
  • Made in the USA
  • Certificates of Conformance sent with shipment
Specifications
Autoclavable Autoclavable
Certifications/Compliance COC
Closure Type Safe Seal
Color Clear, White
Disposable Single-use
For Use With (Application) Steam Sterilization of Pharmaceutical Equipment
Material Tyvek
Sterility Non-sterile
Thickness (Metric) 5.3 mil
Latex Free Yes
Size 14 x 40
Temperature Range (Metric) 121 to 125 C
Thickness (English) 5.3 mil
Length (English) 40 in.
Length (Metric) 101.6 cm
Width (English) 14 in.
Width (Metric) 35.6 cm
Product Type Autoclave Bag