Learn More
Pure Water Density Standard; UKAS ISO/IEC17025 and ISO 17034 certified, nominal density values: 0.9982 g/mL (20°C),0.9970 g/mL (25°C), Triple pack (3 x 60mL), Paragon Scientific™
Product Code. 16080100
Change view
Click to view available options
Quantity:
2 x 60 mL
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16080100 2 x 60 mL Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16080100 Supplier Paragon Scientific Supplier No. DENWAT3
Paragon ISO 17025 / ISO 17034 pure water Density standard is designed for the verification of instruments used to measure density. Typically used for the verification of digital density meters. ISO 17025 / ISO 17034 certification under UKAS ensures full traceability to national standards.
  • Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Certified in accordance with primary level ASTM D1480 methodology
  • The reagent water meets the definition of Type II in the Specification ASTM D1193
  • Supplied in tamper evident packaging with a 12 month shelf life
  • Available as single bottle packs or triple pack kits
  • Manufactured in the United Kingdom
Specifications
Quantity 2 x 60 mL
Nominal values quoted. Certified Values will vary from batch to batch.