Learn More
Buerkle™ Pump Station for AccuOne/EnergyOne Pump
Product Code. 11941581 Shop All Bürkle Products
Change view
Click to view available options
Depth (Metric):
50 cm
70 cm
100 cm
Unit Size:
Each
Product Code. Depth (Metric) unitSize
11941581 100 cm Each
11991591 50 cm Each
11901601 70 cm Each
3 options
Product Code. 11941581
Brand: Buerkle™ 59003100
Only null left
Add to basket
Add to basket
To aid in transferring small quantities of watery media up to 300 mPas from canisters, barrels and narrow-necked containers from 32mm opening diameter.
  • Pump station accessory only
  • Available for three different submersible lengths
TRUSTED_SUSTAINABILITY
Specifications
Depth (Metric) 100 cm
No. per Pack 1 Ea.
Type Pump Station
Applications Dispensing acids and bases
For Use With AccuOne/EnergyOne Pump