Learn More
HUBER Pump Adapter
Includes hose connector NW12
Brand:  HUBER 19606
Additional Details : Weight : 0.80000kg
Product Code. 10527614
  • 1929.20 NOK / Each
Estimated Shipment: 27-06-2024
Description
Pump adapter CPL for KISS E, CC-E and with baths 106A to 118A
Specifications
Specifications
KISS E, CC-E and with Baths 106 A to 118 A
1 Ea.
Pump Adapters