Learn More
PUM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11586-1-AP-150UL
This item is not returnable.
View return policy
Description
LZTS2 is a negative regulator of katanin-mediated microtubule severing and release from the centrosome. LZTS2 is required for central spindle formation and the completion of cytokinesis.Specifications
| PUM2 | |
| Polyclonal | |
| Unconjugated | |
| PUM2 | |
| KIAA0235, PUM2, PUMH2, Pumilio 2, Pumilio homolog 2, pumilio homolog 2 (Drosophila), PUML2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23369, 298874, 80913 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q80U58, Q8TB72 | |
| Pum2 | |
| PUM2 Fusion Protein Ag2179 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |