missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PUM1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26256-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the PUF family, evolutionarily conserved RNA-binding proteins related to the Pumilio proteins of Drosophila and the fem-3 mRNA binding factor proteins of C. elegans. The encoded protein contains a sequence-specific RNA binding domain comprised of eight repeats and N- and C-terminal flanking regions, and serves as a translational regulator of specific mRNAs by binding to their 3′ untranslated regions. The evolutionarily conserved function of the encoded protein in invertebrates and lower vertebrates suggests that the human protein may be involved in translational regulation of embryogenesis, and cell development and differentiation. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
PUM1 | |
Polyclonal | |
Unconjugated | |
PUM1 | |
HSPUM, KIAA0099, PUM1, PUMH, PUMH1, Pumilio 1, Pumilio homolog 1, pumilio homolog 1 (Drosophila), PUML1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9698 | |
-20°C | |
Liquid |
Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q14671 | |
PUM1 | |
PUM1 Fusion Protein Ag24651 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |