missing translation for 'onlineSavingsMsg'
Learn More

PTPRF Mouse anti-Human, Clone: 2D12G7, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 68070-1-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 17639746

  • 3828.53 NOK / 150µL
Estimated Shipment: 02-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

PTPRF Monoclonal antibody apecifically detetects PTPRF antigen in Human Samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

PTPRF
Monoclonal
1000 μg/mL
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
PTPRF
PTPRF fusion protein Ag25441
150 μL
5792
Store at -20°C. Stable for one year after shipment.
IgG1
Western Blot, ELISA
2D12G7
Unconjugated
BC048768
Mouse
Protein G purification
Primary
Human
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers