missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTHLH Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10817-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the parathyroid hormone family. This hormone regulates endochondral bone development and epithelial-mesenchymal interactions during the formation of the mammary glands and teeth. This hormone is involved in lactation possibly by regulating the mobilization and transfer of calcium to the milk. The receptor of this hormone, PTHR1, is responsible for most cases of humoral hypercalcemia of maligcy. Four alternatively spliced transcript variants encoding two distinct isoforms have been observed. There is also evidence for alternative translation initiation from non-AUG (CUG and GUG) start sites, in-frame and downstream of the initiator AUG codon, to give rise to nuclear forms of this hormone.Specifications
| PTHLH | |
| Polyclonal | |
| Unconjugated | |
| PTHLH | |
| HHM, PLP, PTH rP, PTHLH, PTHR, PTHRP, PTHrP[107 139] | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5744 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P12272 | |
| PTHLH | |
| PTHLH Fusion Protein Ag1133 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction