missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTH Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19001-1-AP-20UL
This item is not returnable.
View return policy
Description
PTH is a hormone producted by the parathyroid gland that regulates the concentration of calcium and phosphorus in extracellular fluid. It is produced in the parathyroid gland as an 84 amino acid single chain polypeptide. Defects in this gene are a cause of familial isolated hypoparathyroidism.Specifications
| PTH | |
| Polyclonal | |
| Unconjugated | |
| PTH | |
| Parathormone, Parathyrin, parathyroid hormone, PTH, PTH1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5741 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P01270 | |
| PTH | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction