missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTH Antibody (3H9 + PTH/1175), Alexa Fluor™ 750 - C-terminus, N-terminal, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47731AF750
This item is not returnable.
View return policy
Description
PTH Monoclonal specifically detects PTH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PTH | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PTH | |
| A synthetic peptide around aa 1-34 of human mature-PTH-polypeptide (exact sequence is proprietary) (3H9); A recombinant fragment around aa 32-115 of human mature PTH-polypeptide (exact sequence is proprietary) (Uniprot: P01270) | |
| Primary | |
| Epitope of this monoclonal antibody maps in the C-terminus of PTH, a hormone produced by the parathyroid gland that regulates the concentration of calcium and phosphorus in extracellular fluid. This hormone elevates blood Ca2+ levels by dissolving the salts in bone and preventing their renal excretion. It is produced in the parathyroid gland as an 84 amino acid single chain polypeptide. It can also be secreted as N-terminal truncated fragments or C-terminal fragments after intracellular degradation, as in case of hypercalcemia. Defects in this gene are a cause of familial isolated hypoparathyroidism (FIH); also called autosomal dominant hypoparathyroidism or autosomal dominant hypocalcemia. FIH is characterized by hypocalcemia and hyperphosphatemia due to inadequate secretion of parathyroid hormone. Symptoms are seizures, tetany and cramps. FIH exist both as autosomal dominant and recessive forms of hypoparathyroidism. | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 3H9 + PTH/1175 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence | |
| Parathormone, Parathyrin, parathyroid hormone, parathyroid hormone 1, PTH1 | |
| Mouse | |
| 0.1 mL | |
| 5741 | |
| Human | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction