Learn More
PTGS1 Mouse anti-Human, Unconjugated, Clone: 1F12G7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67346-1-IG-150UL
This item is not returnable.
View return policy
Description
The PTGS1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PTGS1. This antibody recognizes human antigen. The PTGS1 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| PTGS1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunofluorescence, ELISA | |
| 1F12G7 | |
| Unconjugated | |
| 150 μL | |
| 5742 | |
| IgG2b |