Learn More
BRAND™ PTFE Watch Glass
Product Code. 10425651 Shop All Brand Products
Change view
Click to view available options
Diameter (Metric):
40 mm
65 mm
75 mm
125 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
10425651 40 mm Each
10252211 65 mm Each
10303771 75 mm Each
10088490 125 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10425651 Supplier BRAND™ Supplier No. 150904
  • ONLINE EXCLUSIVE PRICE
    120.00 NOK
    116.18 NOK / Pack of 1
    Save 3.82 NOK
    3% Off
Estimated Shipment: 16-12-2026
PTFE Watch Glasses are ideally suited for covering beakers.
  • Constructed of PTFE
  • Ideal for covering PTFE beakers
  • Excellent chemical resistance
TRUSTED_SUSTAINABILITY
Specifications
Material PTFE
Type Watch Glass
For Use With (Equipment) Beaker
Diameter (Metric) 40 mm