Learn More
Thermo Scientific Chemicals PTFE Tweezers-Forceps
Product Code. 15431487
Change view
Click to view available options
Length (Metric):
100 mm
200 mm
Tip Style:
Sharp
Square
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Tip Style unitSize
15431487 200 mm Sharp Each
15406195 100 mm Square Each
15416195 200 mm Square Each
15421487 100 mm Sharp Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15431487 Supplier Thermo Scientific Chemicals Supplier No. 038263.KT
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Totally inert and non-contaminating
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • PTFE
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tweezers-Forceps
Material PTFE
Tip Style Sharp
Length (English) 7.8 in.
Length (Metric) 200 mm