Learn More
Thermo Scientific Chemicals PTFE Tweezers-Forceps
Product Code. 15406195
Click to view available options
Length (Metric):
100 mm
200 mm
Tip Style:
Sharp
Square
Unit Size:
Each
Product Code. 15406195
Brand: Thermo Scientific Chemicals 038260.KT
Request Bulk or Custom Format
Totally inert and non-contaminating
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • PTFE
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tweezers-Forceps
Material PTFE
Tip Style Square
Length (English) 3.9 in.
Length (Metric) 100 mm