Learn More
Saint Gobain Life Sciences PTFE Tubing
Click to view available options
Diameter (Metric) Inner:
0.89 mm
1.6 mm
2 mm
5 mm
9.5 mm
16 mm
Diameter (Metric) Outer:
2.57 mm
3.2 mm
4 mm
7 mm
13 mm
19.6 mm
Unit Size:
10m
1m
25m
50m
Pack of 12
Description
- Chemically inert tubing
- Excellent chemical resistance
- Will not leach into or absorb out of fluid being pumped
- Extremely low gas permeability
- Nontoxic
- Virtually nonporous
- Low coefficient of friction
- Sterilize by EtO or autoclave
Specifications
Specifications
| Autoclavable | Autoclavable |
| Diameter (English) Inner | 0.06 in. |
| Diameter (Metric) Inner | 1.6 mm |
| Material | PTFE |
| Diameter (English) Outer | 0.13 in. |
| Diameter (Metric) Outer | 3.2 mm |
| Overall Length (English) | 32.8 ft. |
| Overall Length (Metric) | 10 m |
| Thickness (English) | 1/32 in. |
| Thickness (Metric) | 0.8 mm |
| Show More |