Learn More
Hirschmann™ PTFE Thread Adapter
Description
- For the adaption on the most common bottles
Specifications
Specifications
| Product Type | Thread Adapter |
| Material | PTFE |
| For Use With (Equipment) | ceramus™ classic and EM-dispenser |
| Quantity | 1 Ea. |
| Thread Size | 45 mm |
| Diameter (Metric) Neck | 32 mm |