Learn More
BRAND™ PTFE Stirring Rod
Product Code. 10657661 Shop All Brand Products
Change view
Click to view available options
Length (Metric):
200 mm
250 mm
300 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
10657661 300 mm Each
10185390 200 mm Each
10370091 250 mm Each
3 options
Product Code. 10657661
Brand: BRAND™ 136225
Only null left
Add to basket
Add to basket
PTFE construction. Stirring rod includes spatulas at each end.
  • PTFE construction
  • With spatulate ends
TRUSTED_SUSTAINABILITY
Specifications
Includes Spatulate ends
Diameter (Metric) 8 mm
Length (Metric) 300 mm
Material PTFE
Quantity 1