Learn More
Thermo Scientific™ PTFE/Silicone Discs
Product Code. 11527340 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric) Outer:
8 mm
12 mm
13 mm
16 mm
18 mm
22 mm
Unit Size:
Pack of 72
Product Code. Diameter (Metric) Outer unitSize
11527340 22 mm Pack of 72
11591004 8 mm Pack of 72
11501014 12 mm Pack of 72
11587330 13 mm Pack of 72
11597330 16 mm Pack of 72
11531014 18 mm Pack of 72
6 options
Product Code. 11527340
Brand: Thermo Scientific™ TS12722
Only null left
Add to basket
Add to basket
Combine the inertness of a PTFE coating with the resealability of silicone Thermo Scientific™ PTFE/Silicone Discs.
Ensure air-tight, inert seals with autoclavable Thermo Scientific™ PTFE/Silicone Discs. They are specifically designed to combine the resealability of silicone with the inertness of PTFE. These discs reseal instantly puncture after puncture, and the PTFE coating is structurally bonded so that no cement is leached or baked out of your sample after needle penetration. Many sizes are available to fit Thermo Scientific™ Reacti-Vial™ Small Reaction Vials, Thermo Scientific™ Screw Cap Septum Vials, and Thermo Scientific™ Hypo-ViaI™ Sample Storage Vials.
Additional Product Information
Each disc compresses to maintain a tight seal, forcing the PTFE coating to conform to the sealing surface.
Standard syringe and CC needles penetrate an entire disc with ease.
The PTFE layer is a full 10 mils thick to ensure protection
TRUSTED_SUSTAINABILITY
Specifications
Description Diameter: 22mm
Diameter (Metric) Outer 22 mm
Material PTFE, Silicone
Quantity 72 Pk.
Thickness (Metric) 10 mil
For Use With (Equipment) 25 and 40 mL Screw Cap Septum Vial, 240 mL (8 oz.) Screw Cap Bottle, 10 mL Reacti-Vial
Autoclavable Autoclavable
Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC)
Product Type Disc