missing translation for 'onlineSavingsMsg'
Learn More

PTFE sheet, 12.7mm (0.5in) thick

Product Code. 15462207
Click to view available options
Quantity:
300 x 300 mm
Packaging:
Shrink-wrapped
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15462207

Brand: Thermo Scientific Alfa Aesar 045213.HB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Shrink-wrapped
MDL Number MFCD00084455
Quantity 300 x 300 mm
Chemical Name or Material PTFE sheet

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.