Learn More
Bochem™ Sharp Tip Forceps with Guide Pin
With a guide pin to help keep the tips aligned.
Brand:  Bochem™ 1812
 View more versions of this product
Product Code. 10587882
  • 378.00 NOK / Each
Only null left
Estimated Shipment: 26-09-2025
Description
  • Manufactued from PTFE-coated stainless steel
  • Sharp tips for excellent precision
Specifications
Specifications
Sharp
Stainless Steel
Forceps
Guide pin
1 Pack
145 mm
PTFE coating
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bochem™ Sharp Tip Forceps with Guide Pin > Length: 145mm