Learn More
Cole-Parmer™ PTFE Screen Gaskets
Product Code. 16659285
Click to view available options
Diameter (Metric) Outside:
1.27 cm
2.54 cm
3.81 cm
5.08 cm
Mesh Size:
4
10
20
40
100
Unit Size:
Each
Product Code. 16659285
Brand: Cole-Parmer™ 3053844
Only null left
Add to basket
Add to basket
Protect your pump, valves and other components in fluid line
  • Pre-filter before critical membrane filters
  • Generate turbulence for enhanced mass or heat transfer
  • Purify in chromatography applications
Specifications
Mesh Size 4
Diameter (Metric) Outside 2.54 cm
Material PTFE
Type Gasket