Learn More
Cole-Parmer™ PTFE Screen Gaskets
Click to view available options
Diameter (Metric) Outside:
1.27 cm
2.54 cm
3.81 cm
5.08 cm
Mesh Size:
4
10
20
40
100
Unit Size:
Each
Description
- Pre-filter before critical membrane filters
- Generate turbulence for enhanced mass or heat transfer
- Purify in chromatography applications
Specifications
Specifications
| Mesh Size | 100 |
| Diameter (Metric) Outside | 3.81 cm |
| Material | PTFE |
| Type | Gasket |